Learn More
Grainger Flex Metal Conduit Steel Trd Sz 1/2in
Supplier:  Grainger 16R040
Encompass_Preferred
The AFC Liquid Tight Flex Metal Conduit is a flexible metal conduit with a liquid tight PVC jacket. It is made of galvanized steel and has an operating temperature range of -20°F to 167°F. The conduit has a 3/4" trade size, is sold by the foot, and is UL and CSA listed. It can be used to protect and route electrical wiring in damp, wet location applications both indoors and outdoors. The conduit is sunlight, oil, and chemical resistant. It meets NEC code requirements for liquid tight flexible metal conduit.
Catalog No. 50-276-5177
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.