Learn More
Sigma Aldrich Fine Chemicals Biosciences Worthington LD Series Liqu1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F4524
Encompass
LD Series Liquid Nitrogen Dewars are designed for storage of small quantities of liquid nitrogen. High-strength aluminum shell is both durable and light-weight. Super insulation provides advanced thermal protection. Neck tube joins inner and outer container providing maximum durability and minimum nitrogen consumption. Cap and neck tube core provide positive closure.
Catalog No. 50-290-8985
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.