Learn More
Enzo Life Sciences Wortmannin (5 mg). CAS: 19545-26-7
SDP
Supplier:  Enzo Life Sciences BMLST4150005
Encompass_Preferred
Potent and selective inhibitor of phosphatidyl-inositol 3-kinase. Active in purified preparations and cytosolic fractions(IC50 = 5nM) and is highly cell permeable. Inhibits fMLP induced PIP3 and superoxide anion production (IC50 = 50nM) in guinea pig neutrophils and blocks the metabolic effects of insulin in isolated rat adipocytes without affecting the insulin receptor tyrosine kinase activity. Covalently binds to PI 3-kinase and is selective, inhibiting other kinases such as PI 4-kinase and myosin light chain kinase at concentrations 100-fold higher than that required for inhibition of PI 3-kinase. The half-life of wortmannin in culture media is quite short (8-13 minutes), care should be taken to account for this decomposition when planning experiments. Alternative name: KY 12420. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in DMSO (50mg/ml), methanol (5mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2181
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.