Learn More
Ampco Safety Tools Wrench, Hex Key
SDP
Catalog No. 19174765 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Size:
1/16 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
5/64 in.
7/32 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-174-765 3/16 in.
19-174-751 1/16 in.
19-174-752 1/8 in.
19-174-766 3/32 in.
19-174-772 5/32 in.
19-174-773 5/64 in.
19-174-776 7/32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.; WARNING: Cancer - www.P65Warnings.ca.gov
Catalog No. 19-174-765 Supplier Ampco Safety Tools Supplier No. WH3/16
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.; WARNING: Cancer - www.P65Warnings.ca.gov
AMPCO™Hex Key Wrench performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Hex Key
Material Beryllium Copper
Size 3/16 in.
Certifications/Compliance Factory Mutual (FM) Approved
WARNING: Cancer - www.P65Warnings.ca.gov
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.