Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wright's Stain, Regular Formula, for Staining and General Differentiation of Blood Corpuscles, Ricca Chemical
SDP

Catalog No. 93601 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
93601 4 L Amber Glass Bottle
936016 500 mL Amber Glass Bottle
936032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 93601 Supplier Ricca Chemical Company Supplier No. 93601
Add to Cart
Add to Cart

Staining and General Differentiation of Blood Corpuscles

Specifications

Chemical Name or Material Wright's Stain
Name Note Regular Formula
CAS 67-56-1, 68988-92-1, 7558-79-4, 7778-77-0, 7664-38-2, 7732-18-5
CAS Min % 97.0, 0.38, 0.08, 0.04, 0.0010
CAS Max % 100.0, 0.39, 0.09, 0.04, 0.0010
Grade Laboratory
Packaging Amber Glass Bottle
Color Dark Purple
pH 6.30 to 6.50
Quantity 4 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.