Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wright' Stain (Powder/Certified Biological Stain), Fisher Chemical™

Catalog No. W35100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
W35-100 100 g Glass Bottle
W35-25 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. W35-100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

For use in blood staining

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68988-92-1

Specifications

Bio. Stain Comm. Certification Pass Test
Molecular Formula C36H27Br4N3O5S+2
MDL Number MFCD00082143
Identification Pass Test
InChI Key AXIKDPDWFVPGOD-UHFFFAOYSA-O
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid
Molecular Weight (g/mol) 933.304
PubChem CID 25113599
Grade Certified Biological Stain
Packaging Glass Bottle
Color Green
Quantity 100 g
Physical Form Solid
Chemical Name or Material Wright«s Stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.