Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WSB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562650 Supplier Invitrogen™ Supplier No. PA562650
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVTASYDTNV IMWDPYTGER LRSLHHTQVD PAMDDSDVHI SSLRSVCFSP EGLYLATVAD DRLL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

This gene encodes a member of the WD-protein subfamily. The encoded protein contains five WD-repeats spanning most of the protein and an SOCS box in the C-terminus. The SOCS box may act as a bridge between specific substrate-binding domains and E3 ubiquitin protein ligases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WSB2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WSB2
Gene Accession No. Q9NYS7
Gene Alias AA673511; CS box-containing WD protein; SBA2; SOCS box-containing WD protein SWiP-2; Swip2; WD repeat and SOCS box containing 2; WD repeat and SOCS box containing protein 2; WD repeat and SOCS box-containing 2; WD repeat and SOCS box-containing protein 2; WD SOCS-box protein 2; WD-40-repeat-containing protein with a SOCS box 2; Wsb2; WSB-2
Gene Symbols WSB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WSB2. Recombinant protein control fragment (Product #RP-101513).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55884
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.