Learn More
Cayman Chemical WST-1 Cell Proliferation Assay Kit, 480 wells
SDP
Supplier:  Cayman Chemical 10008883480WELLS
Encompass_Preferred
Cayman’s WST-1 Proliferation Assay is based on the reduction of tetrazolium salt WST-1 to a soluble formazan dye by electron transport across the plasma membrane of dividing cells. This kit will also allow investigators to screen drug candidates involved in cell cycle regulation.
Catalog No. NC0282500
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.