Learn More
Cayman Chemical WST-8 Cell Proliferation Assay Kit, 480 wells
Supplier: Cayman Chemical 10010199480WELLS
Cayman's WST-8 Cell Proliferation Assay provides a tool for studying induction and inhibition of cell proliferation in any in vitro model. The assay is based on the extracellular reduction of WST-8 by NADH produced in the mitochondria resulting in a water-soluble formazan which dissolves directly into the culture medium. Cayman's WST-8 assay is preferred when higher cell densities are expected, as up to 5 x 106 cells/ml can be successfully quantified.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.