Learn More
ATCC WU E-4 [1506 A1.4] hybridoma producing monoclonal anti-human apolipoprotein E (IgG1) - ATCC CRL-2247
Supplier: ATCC CRL2247
WU E-4 [1506 A1.4] is a mouse hybridoma cell line that produces a monoclonal IgG1 antibody specific for human apolipoprotein E (ApoE). Derived by fusion of mouse spleen B cells with Sp2/0-Ag14 myeloma cells, the antibody recognizes an epitope within amino acids 126-191 of ApoE and can block ApoE-mediated binding of lipoproteins to the LDL receptor. For research use only.
- Produces monoclonal IgG1 antibody specific for human apolipoprotein E (ApoE).
- Derived from fusion of mouse spleen B cells with Sp2/0-Ag14 myeloma cells.
- Complete medium: DMEM (4.5 g/L glucose, 0.1 mM non-essential amino acids, 1 mM sodium pyruvate) supplemented with 10% horse serum and 5% fetal bovine serum.
- Culture conditions: 37°C, 95% air, 5% CO2.
- Storage: vapor phase of liquid nitrogen; cryopreserve in complete medium with 5% (v/v) DMSO.
- Mycoplasma: not detected.
- Tumorigenic: forms ascites in pristane-primed BALB/c mice.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.