Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WWP1 Antibody [Alexa Fluor Plus 405], R&D Systems™
SDP

Catalog No. RD10020536
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10020536 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10020536 Supplier R&D Systems Supplier No. NBP149712AFP405

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

WWP1 Polyclonal antibody specifically detects WWP1 in Human samples. It is validated for Immunoprecipitation,Western Blot

Specifications

Antigen WWP1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor Plus 405
Formulation 50mM Sodium Borate
Gene Alias AIP5Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 5, Atrophin-1-interacting protein 5, DKFZP434D2111, EC 6.3.2, EC 6.3.2.-, hSDRP1, NEDD4-like E3 ubiquitin-protein ligase WWP1, TGIF-interacting ubiquitin ligase 1, TIUL1, WW domain containing E3 ubiquitin protein ligase 1, WW domain-containing protein 1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 200 and 250 of human WW Domain Containing E3 Ubiquitin Protein Ligase 1 using the numbering given in entry NP_008944.1 (GeneID 11059).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11059
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.