Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam WY 14643, PPARalpha Agonist, >99% Purity, Solid, 50 mg
Description
- Selective PPARα agonist: EC50 values are 0.63, 32 and >100 μM at PPARα, PPARγ and PPARδ receptors respectively.
- Purity: >99%
- Molecular weight: 323.8 g/mol
- Form: Solid
- Anti-inflammatory: Able to reduce inflammatory responses in vivo and in vitro.
- CAS Number: 50892-23-4
- Storage: Store at Ambient.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | PPARα (PPARγ, PPARδ) |
| Shipping Condition | Ambient |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% ((4-Chloro-6-(2,3-xylidino)-2-pyrimidinyl)thio)acetic acid |
| Form | Solid |
| Formulation | 100% ((4-Chloro-6-(2,3-xylidino)-2-pyrimidinyl)thio)acetic acid |
| Quantity | 50 mg |
| For Use With (Application) | PPARalpha agonist, inflammatory response modulation |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?