Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam WY 14643, PPARalpha Agonist, Purity >99%, Solid, 10 mg

Catalog No. 502706853
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6853 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6853 Supplier Abcam Supplier No. AB14114210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly selective PPARα agonist capable of reducing inflammatory responses.

  • Molecular Weight: 323.8 g/mol
  • Purity: >99%
  • Form: Solid
  • Activity: Selective PPARα agonist (EC50 values are 0.63, 32 and >100 μM at PPARα, PPARγ and PPARδ receptors respectively)
  • Biological Function: Reduces inflammatory responses in vivo and in vitro
  • CAS Number: 50892-23-4
  • Storage Temperature: Store at Ambient

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule PPARα
Form Solid
Buffer 100% ((4-Chloro-6-(2,3-xylidino)-2-pyrimidinyl)thio)acetic acid
Content And Storage Store at ambient temperature.
Formulation 100% ((4-Chloro-6-(2,3-xylidino)-2-pyrimidinyl)thio)acetic acid
Quantity 10 mg
For Use With (Application) PPARalpha agonist
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.