Learn More
Apexbio Technology LLC WZ4002, 10mg. Cas: 1213269-23-8 MFCD: MFCD16621244. Mutant-selective EGFR inhibitor(L858R,T790M), irreversible and potent
SDP
Supplier:  Apexbio Technology LLC A138910
Encompass_Preferred
WZ4002 is a novel mutant-selective epidermal growth factor receptor (EGFR) kinase inhibitor that potently and irreversibly suppresses the ATP-dependent auto-phosphorylation of both wild-type EGFR and EGFR mutants. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4696
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.