Learn More
Selleck Chemical LLC WZB117 10mg 1223397-11-2
SDP
Supplier:  Selleck Chemical LLC S792710MG
Encompass_Preferred
WZB117 is an inhibitor of Glucose Transporter 1 (GLUT1). It inhibited cell proliferation in lung cancer A549 cells and breast cancer MCF7 cells with an IC50 of approximately 10 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5425