Learn More
Description
- Selected cells are immediately available for downstream applications such as flow cytometry and cell culture expansion.
- Easy to use
- Single-use, column-free and gentle on cells
- High purity: Achieve purities of up to 99%
- Reproducible results (>86%)
- High viability (>97%)
- Process up to 4 x 108 total cells
Specifications
Specifications
| Capacity (Metric) | Up to 4 x 108 total cells |
| For Use With (Application) | Blood and Blood Products |
| Quantity | 1/Pk., 1/Cs. |
| Content And Storage | 2°C to 8°C |
| Includes | Biotin Mouse Anti-Human Antibody; Corning X-BACS™ Reagent; Corning X-BACS Buffer; Syringe, 10 mL; 21 G x 1.5 in. Needle; Syringe cap |
| Label Type | Biotinylated anti-human CD3 Antibody |
| Product Type | Cell selection |
| Detection Method | Buoyancy Activated Cell Sorting |
| Product Line | Corning X-BACS |
| Shipping Condition | Room Temperature |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.