Learn More
3M™ X.Sport™ Eyewear
Ideal choice for active wear
Supplier: 3M™ 7000127553
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
- Sport-grip™ temples
- Ratchet adjustment for three lens angles
- 7.5 base lens is mounted in a lightweight nylon frame
- Provides maximum coverage and excellent peripheral vision
- Lens with DX™ anti-fog hard coat protects from fog, scratching, and most chemicals
- 3M Catalog# 15176-00000-20 , 3M Catalog# 15182-00000-20
Compliance
Meets the requirements of ANSI™ Z87.1-2010 and CSA Z94.3-2007.
Specifications
Anti-fog | |
Clear | |
15176-00000-20 | |
Nylon |
Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007. | |
Black | |
Sport Grip |