Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ X-Y Axis Dry Erase Mat
SDPSCI_ED

Catalog No. S111005 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
10 Pk.
30 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S111005 10 Pk.
S01305 30 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Prop 65
Catalog No. S111005 Supplier United Scientific™ Supplier No. UNCG9X11PK/10
  • $10.25
    $9.65 / Pack of 10

    Save $0.60

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65

Double sided dry-erase mat includes a 1 cm x-y axis grid on one side for graphing activities.

  • Double sided dry-erase mat
  • Includes a 1 cm x-y axis grid on one side for graphing activities.
  • The other side is blank but coated so it can be used with dry-erase markers.

Specifications

Product Type Dry Erase Board
For Use With (Application) Graphing
Quantity 10 Pk.
Dimensions (L x W) 9 x 11 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.