Learn More
Montana Molecular Green Gi cADDis cAMP Sensor Kit X0200G, 60mL, CMV
SDP
Supplier:  Montana Molecular X0200G060
Encompass_Preferred
Measure cyclic AMP (cAMP) signaling kinetics, screen GPCRs for Gi activity and more with this live cell fluorescent assay. Add cADDis to your cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. Decreasing green fluorescence indicates cAMP increase. The assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. Kits include positive control receptors and agonists.
Catalog No. 50-163-1910
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.