Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ XAC
GREENER_CHOICE

Catalog No. 320010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-001-0 10 mg
32-005-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-001-0 Supplier Tocris Bioscience™ Supplier No. 3200/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Adenosine receptor antagonist

Adenosine receptor antagonist (IC50 values are 1.8 and 114 nM at A1 and A2 receptors respectively). Attenuates adenosine-induced vasodilation and exhibits proconvulsant activity in vivo.

Chemical Identifiers

CAS 1962928-23-9
Molecular Formula C21H30Cl2N6O4

Specifications

Quantity 10 mg
Formula Weight 464.95
Purity >98%
Molecular Weight (g/mol) 501.41
Chemical Name or Material XAC
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.