Learn More
Invitrogen™ XAF1 Polyclonal Antibody

Description
Suggested positive control: antigen standard for XAF1 (transient overexpression lysate), human liver tissue or protein. Immunogen sequence: NQTSTMEKDV RPKTRSIN & QLDLEKERY YKFKRFHF.
Specifications
Specifications
| Antigen | XAF1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | XAF1 |
| Gene Accession No. | Q5NBU8, Q6GPH4 |
| Gene Alias | BIRC4 binding protein; BIRC4-binding protein; Birc4bp; F-box protein 39; Fbox39; HSXIAPAF1; novel TRAF-type zinc finger protein; XAF1; XIAP associated factor 1; XIAP associated factor-1; Xiapaf1; XIAP-associated factor 1 |
| Gene Symbols | XAF1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.