Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthurenic Acid 96.0+%, TCI America™
SDP

Catalog No. X00545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00545G 5 g
X00541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00545G Supplier TCI America Supplier No. X00545G

Chemical Identifiers

CAS 59-00-7
Molecular Formula C10H7NO4
Molecular Weight (g/mol) 205.17
MDL Number MFCD00006754
InChI Key FBZONXHGGPHHIY-UHFFFAOYSA-N
Synonym xanthurenic acid, 4,8-dihydroxyquinoline-2-carboxylic acid, xanthuric acid, 8-hydroxykynurenic acid, 4,8-dihydroxyquinaldic acid, 4,8-dihydroxyquinaldinic acid, xanthurenate, 2-quinolinecarboxylic acid, 4,8-dihydroxy, unii-58lab1bg8j, ccris 4429
PubChem CID 5699
ChEBI CHEBI:10072
IUPAC Name 8-hydroxy-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C2=CC=CC(O)=C2N1

Specifications

Quantity 5 g
Formula Weight 205.17
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Xanthurenic Acid
RTECSNumber UZ9275000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.