Learn More
WELCH MATERIALS INC Ultisil XB-C18, 10µm, 300Å, 50×250mm
Supplier:  WELCH MATERIALS INC H0261032210
Encompass_Preferred
Prep Column, Ultisil XB-C18, 10µm, 300Å, 50×250mm, pH range: 1.5-10.0, Pore Size: 300Å, USP listing: L1, Carbon Loading: 8%, Surface Area: 90m2/g, endcapped.
Catalog No. NC2052592
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.