Learn More
Waters Corp XBridge Peptide BEH C18 Column, 300Å, 10 µm, 4.6 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186003665
Encompass_Preferred
The XBridge BEH300 C18 Peptide Separation Technology columns are optimized and QC tested for peptide separations. The wide-pore (300Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
Catalog No. 50-781-937
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.