Learn More
Waters Corp XBridge Protein BEH C4 OBD Prep Column, 300Å, 5 µm, 19 mm X 250 mm, 1/pk
Supplier:  Waters Corp 186007314
Encompass_Preferred
Waters Protein Separation Technology columns are QC tested and optimized to separate proteins based on size, hydrophobicity and isoelectric point. If you compare traditional to C18 phases, the C4 ligand is less retentive and will minimize protein carryover, increase protein recovery and improve peak capacity.
Catalog No. NC1554391
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.