Learn More
Waters Corp XBridge Protein BEH SEC Column, 200Å, 3.5 µm, 7.8 mm X 300 mm, 1/pk
Supplier:  Waters Corp 186007640
Encompass_Preferred
XBridge Protein BEH SEC, 125Å; Columns, containing 3.5 µm particles are based on the same Waters Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating as used in our successful line of HPLC-based SEC columns. They are best suited to separate proteins ranging from 1,000 ; 80,000 Daltons. Note: P/N 176003593 contains an XBridge Protein BEH SEC, 125 Å; 3.5 µm; 7.8 x 300 Column with one vial the BEH125 SEC Protein Std (P/N 186006519)
Catalog No. NC1493851
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.