Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROFLEX™ XCEED™ XC-310 Nitrile Gloves
SCI_ED

Catalog No. S28985 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S28985 X-Large
S28981 X-Small
S28982 Small
S28983 Medium
S28984 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S28985 Supplier Ansell Supplier No. XC310XL/EMD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thin, powder free nitrile exam glove with ERGOFORM™ is designed to reduce hand fatigue.

  • Lightweight and comfortable with less than 3 mil palm thickness
  • Reduce hand fatigue and help workers be more productive while exerting less muscle effort
  • Contains no natural rubber latex or powder to help protect wearers from skin allergies, irritation, and dryness
  • Beaded cuff
  • Textured fingers
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Thickness (English) 2.8 mil
Thickness (Metric) 0.07 mm
Model XC-310
Length (Metric) 24 cm

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.