Learn More
StatLab XCEL PLUS FIXATIVE
SDP
Supplier:  StatLab FXEXPGAL
Encompass_Preferred
Excell Plus is a pleasant smelling, low-hazard tissue fixative that works just like 10% formalin. It is non-flammable, contains no heavy metals and is virucidal, biocidal, and fungicidal.
Catalog No. NC0498416
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.