Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies™ xCELLigence RTCA DP Bundle

Catalog No. 380601050
Change view
Click to view available options
High-throughput Compatibility:
16-well
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. High-throughput Compatibility
38-060-1050 16-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-060-1050 Supplier Agilent Technologies™ Supplier No. 380601050
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

1 xCELLigence RTCA DP instrument, 1 control unit (laptop PC), 3 xCELLigence RTCA Software Pro basic single license keys and 6 xCELLigence RTCA E-Plate 16 PET

Continuously measures cell invasion and migration (CIM) and monitors cell health, behavior, and function using label-free cellular impedance.

  • xCELLigence RTCA DP model can run up to three separate electronic 16-well plates (E-Plate 16 or CIM-Plate 16)
  • Each well in the CIM-Plate 16 is an electronically integrated Boyden chamber, which enables the monitoring of cell invasion and migration in real-time (at second or minute intervals) without exogenous labels
  • Non-invasive and label-free real-time impedance assay using E-plate 16 measures cell health, viability, or response to treatments with high sensitivity, accuracy, and reproducibility
  • Obtain continuous data at high temporal resolution (from seconds to days)
  • Easy workflow allows users to simply add cells to E-plates and begin kinetic measurements at physiological conditions, with minimal hands-on time
  • Intuitive RTCA Software Pro allows for analysis of real-time data to generate diverse plot types and automatically calculate parameters such as % of cytolysis, IC50 or KT50
  • RTCA Software Pro supports FDA 21 CFR Part 11 compliance to ensure electronic data integrity

Specifications

Shaking No
For Use With (Application) Cell Invasion and Migration, Immunotherapy
Detection Method Impedance
High-throughput Compatibility 16-well
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.