Learn More
Apexbio Technology LLC XEN445, 100mg Cas# 1515856-92-4
SDP
Supplier:  Apexbio Technology LLC B4663100
Encompass_Preferred
Endothelial lipase(EL) inhibitor. XEN445 10mg. 195514-63-7. MFCD25976784 XEN445 is a novel, highly selective and potent inhibitor of endothelial lipase(EL) with an IC 50 value of 0.237M [1].In vitro, XEN445 has been reported to selectively inhibit EL with IC50 values of 0.237M, 20M and 9.5M for hEL, hLPL and hHL, respectively. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8707
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.