Learn More
FUJIFILM IRVINE SCIENTIFIC INC DMSOFRE proteNFREE ANIML 100ML
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 91140100ML
Encompass_Preferred
Eliminate the risk of DMSO toxicity and maintain potency of human mesenchymal stem/ stromal cells (MSCs), PBMCs (T cells), and hematopoietic stem cells (HSCs) throughout cryopreservationComparable post-thaw cell viability to solutions containing DMSONontoxic when injected in animal models (refer to toxicological application note)Enables cell preservation for short-term at -80°C* and long-term storage in liquid nitrogen to -196°CComplete, ready-to-use medium* Human MSC and PBMC (T cell) data available for short-term storage. Human HSC data is not available.By purchasing this product you are agree that you will not use this product as excipient
Catalog No. 50-289-2209
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.