Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies™ XFp T Cell Metabolic Profiling Kit

Catalog No. 103771100
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-377-1100 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-377-1100 Supplier Agilent Technologies™ Supplier No. 103771100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XFp T Cell Metabolic Profiling Kit contains six single-use pouches, each with oligomycin A, BAM15, and rotenone/antimycin A. For use with Seahorse XF HS Mini and XFp analyzers.

  • Generates robust bioenergetic data linked to critical T cell attributes for antitumor properties: T cell persistence and T cell metabolic fitness
  • Provides a comprehensive view of T cell and NK cell metabolism, including quantification of both glycolytic and mitochondrial effects
  • Evaluates different construct designs, engineering strategies, starting material selection, or manufacturing conditions
  • Includes improved reagents for more accurate results and minimal optimization of uncouplers for different T and NK cell populations
  • Reconstitute compounds directly in the assay medium, eliminating the need for DMSO
  • Is validated for best performance with the Agilent™ Seahorse XF Pro analyzer, Agilent™ XF HS Mini analyzer, Agilent™ Seahorse XF RPMI medium (pH 7.4), and Agilent™ Seahorse XF supplements
  • Supplies easy-to-interpret data analysis provided by Agilent™ Seahorse Analytics, a cloud-based platform, simplifying data processing

Specifications

Detection Method Fluorescence
For Use With (Equipment) Seahorse Analyzer
Includes Lyophilized Reagents
Quantity 1 Kit
Type Reagent Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.