Learn More
Sigma Aldrich Fine Chemicals Biosciences XIAP Active human recombin
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SRP023020UG
Encompass
XIAP (X-linked inhibitor of apoptosis) is an anti-apoptotic protein belonging to the inhibitor of apoptosis protein (IAP) family an evolutionary conserved protein family. XIAP contains three tandem copies of BIR (baculovirusIAP repeat) domain which is composed of A¼70 residues. IAP family members also contain a RING (really interesting new gene) zinc finger domain in their C-terminals.
Catalog No. 50-290-5421
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.