Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva XK Chromatography Columns (Empty)
GSA_VA

Catalog No. 4520047 Shop All Cytiva Products
Change view
Click to view available options
Height (Metric):
200 mm
300 mm
400 mm
600 mm
700 mm
1000 mm
Diameter (Metric) Inner:
16 mm
26 mm
50 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Diameter (Metric) Inner
45-200-47 200 mm 16 mm
45-200-46 300 mm 50 mm
45-200-48 200 mm 26 mm
45-200-49 400 mm 26 mm
45-200-50 200 mm 50 mm
45-200-53 1000 mm 16 mm
45-200-51 400 mm 16 mm
45-200-52 700 mm 16 mm
45-200-54 700 mm 26 mm
45-200-56 600 mm 50 mm
45-200-55 1000 mm 26 mm
45-200-57 1000 mm 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 45-200-47 Supplier Cytiva Supplier No. 28988937
Add to Cart
Edge
Add to Cart

Ensure trouble-free operation for low to medium pressure. Cytiva XK Chromatography Columns (Empty) connect directly to ÄKTA design instruments or other liquid chromatography systems. The user-friendly ensures reproducible, accurate packing.

XK columns are constructed of robust borosilicate glass, which exhibits excellent chemical resistance and is compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules. The clear tubes allow for visual inspection of the media bed and are available in a broad range of column dimensions for use with many different purification needs.

Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations.

  • Plunger design to ensure a homogeneous buffer distribution and reproducible packing
  • Adapter QuickLock mechanism that facilitates column handling and cleaning
  • Fittings for direct connection to chromatographiy systems
  • Cooling jacket for sensitive purifications
  • Extensive range of spare parts available, facilitating maintenance

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type XK Chromatography Columns (Empty)
Bed Volume Bed height (one adapter): 25 to 155mm
Bed height (two adapters): 0 to 155mm
Diameter (Metric) Inner 16 mm
Height (Metric) 200 mm
Flow Rate Inlet/Outlet (i.d.): 0.75mm
Material Borosilicate glass
Max. Pressure 5 bar
Particle Size 10μm pore size
Support Type Polypropylene (reinforced glass fiber)/Polyamide Net
Column Type Empty Chromatography Column
For Use With (Application) Liquid chromatography at low to medium pressure
Show More Show Less

Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol.

Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.