Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XKR8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546668 Supplier Invitrogen™ Supplier No. PA546668
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CWKPDPDQVD GARSLLSPEG YQLPQNRRMT HLAQKFFPKA KDEAASPVKG Sequence homology: Human: 100%; Rabbit: 86%.

Promotes phosphatidylserine exposure on apoptotic cell surface, possibly by mediating phospholipid scrambling. Phosphatidylserine is a specific marker only present at the surface of apoptotic cells and acts as a specific signal for engulfment. Has no effect on calcium-induced exposure of phosphatidylserine. Activated upon caspase cleavage, suggesting that it does not act prior the onset of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XKR8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene XKR8
Gene Accession No. Q9H6D3
Gene Alias hXkr8; RP11-460I13.3; X Kell blood group precursor-related family, member 8; XK related 8; XK, Kell blood group complex subunit-related family, member 8; XKR8; XK-related protein 8; XK-related protein 8, processed form; X-linked Kx blood group related 8; XRG8
Gene Symbols XKR8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human XKR8 (aa 346-395).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55113
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.