Learn More
Apexbio Technology LLC XL019, 10mg. Cas: 945755-56-6 MFCD: MFCD24386874. JAK2 inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A393710
Encompass_Preferred
XL019 is a potent and selective inhibitor of JAK2 with IC50 value of 2.2 nM.JAK kinases were first identified in 1989 and characterized by the presence of two tyrosine kinase domains. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4848
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.