Learn More
Selleck Chemical LLC XL388
SDP
Supplier:  Selleck Chemical LLC S703510MG
Encompass_Preferred
XL388 is a highly potent selective ATP-competitive inhibitor of mTOR with IC50 of 9 9 nM 1000-fold selectivity over the closely related PI3K kinases
Catalog No. 50-217-3247
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.