Learn More
TARGETMOL CHEMICALS INC XL413 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6735
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. XL413 hydrochloride (BMS-863233 Hydrochloride) is a potent selective and ATP competitive inhibitor of Cdc7 with an IC 50 of 3.4 nM. XL413 hydrochloride also exhibits potent effect with IC 50 s of 215 42 nM on CK2 PIM1 respectively and an EC 50 of 118 nM on pMCM. purity: 99%
Catalog No. 50-257-6468
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.