Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ XLD Agar (Dehydrated)

Catalog No. OXCM469B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM469B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM469B Supplier Thermo Scientific™ Supplier No. CM0469B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate Salmonella and Shigella from food and clinical specimens with this selective medium.

Isolate Salmonella and Shigella from clinical specimens and foods with selective Thermo Scientific™ Oxoid™ XLD Agar (Dehydrated). The medium relies on xylose fermentation, lysine decarboxylation and production of hydrogen sulphide for the primary differentiation of shigellae and salmonellae from non-pathogenic bacteria. The careful addition of sodium desoxycholate inhibits the growth of coliforms without decreasing the ability to support shigellae and salmonellae.

Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.

Use XLD Medium for the screening of samples containing mixed flora suspected of harboring enteric pathogens, e.g., medical specimens or food products.

  • Easy-to-read: easily differentiate Salmonella (red colonies with black centers) from Shigella (red colonies) and other coliforms (yellow, opaque colonies).
  • Superior sensitivity and selectivity: exceeds that of the traditional plating media e.g., Eosin Methylene Blue and Bismuth Sulphite Agars, which tend to suppress the growth of shigellae.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description XLD Medium
Form Powder
Product Type Agar
Quantity 500 g
Yield For 9.4L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.