Learn More
Invitrogen™ XPA Polyclonal Antibody

Description
Positive Control: Hela cell lysate, A431 cell lysate, rat large intestine tissue, human thyroid tissue, human colon carcinoma tissue, human skin tissue, human prostate carcinoma tissue, human breast tissue, human breast carcinoma tissue, human stomach carcinoma tissue, human small intestine tissue, mouse testis tissue, mouse kidney tissue. Subcellular Location: Nucleus.
Specifications
Specifications
| Antigen | XPA |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | XPA |
| Gene Accession No. | P23025, Q64267 |
| Gene Alias | AI573865; DNA repair protein complementing XP-A cells; DNA repair protein complementing XP-A cells homolog; excision repair-controlling; mutant xeroderma pigmentosum complementation group A; xeroderma pigmentosum group A-complementing protein; Xeroderma pigmentosum group A-complementing protein homolog; xeroderma pigmentosum, complementation group A; XP1; XPA; XPA, DNA damage recognition and repair factor; Xpac |
| Gene Symbols | XPA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.