Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA123344
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA123344 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA123344 Supplier Invitrogen™ Supplier No. PA123344
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa Nuclear extract.

The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne's syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2
Gene Ercc3
Gene Accession No. P19447
Gene Alias Basic transcription factor 2 89 kDa subunit; BTF2; BTF2 p89; BTF2-p89; DNA excision repair protein ERCC-3; DNA repair helicase; DNA repair protein complementing XP-B cells; ERCC excision repair 3, TFIIH core complex helicase subunit; Ercc3; Ercc-3; excision repair 3; excision repair cross-complementation group 3; excision repair cross-complementing rodent repair deficiency, complementation group 3; General transcription and DNA repair factor IIH helicase subunit XPB; GTF2H; RAD25; TFIIH; TFIIH 89 kDa subunit; TFIIH basal transcription factor complex 89 kDa subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH p89; TFIIH subunit XPB; TTD2; Xeroderma pigmentosum group B-complementing protein; xeroderma pigmentosum, complementation group B; XPB; XPBC
Gene Symbols Ercc3
Host Species Rabbit
Immunogen Peptide corresponding to the carboxy-terminus of human XPB.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2071
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.