Learn More
Maine Molecular Quality Controls Inc Xpert FII & FV HET
SDP
Catalog No. NC2005274
Encompass
Xpert FII & FV HET is intended for in vitro use as a quality control to monitor analytical performance of FII & FV detection on the GeneXpert System. Xpert FII & FV HET is formulated to monitor the detection of mutations Factor II G20210A and Factor V Leiden G1691A, the most common genetic risk factors for thrombotic events. Factor II and Factor V DNA in a proprietary matrix with preservatives. The kit is comprised of two bottles of Xpert FII & FV Heterozygous Control which contain heterozygous DNA for both Factor II and Factor V. Xpert FII & FV HET is comprised of two bottles containing 0.5mL each of synthetic Factor II and Factor V DNA suspended in a non-infectious matrix with carrier DNA, preservatives and stabilizers.
Catalog No. NC2005274 Supplier Maine Molecular Quality Controls Inc Supplier No. G1082H
Xpert FII & FV HET is intended for in vitro use as a quality control to monitor analytical performance of FII & FV detection on the GeneXpert System. Xpert FII & FV HET is formulated to monitor the detection of mutations Factor II G20210A and Factor V Leiden G1691A, the most common genetic risk factors for thrombotic events. Factor II and Factor V DNA in a proprietary matrix with preservatives. The kit is comprised of two bottles of Xpert FII & FV Heterozygous Control which contain heterozygous DNA for both Factor II and Factor V. Xpert FII & FV HET is comprised of two bottles containing 0.5mL each of synthetic Factor II and Factor V DNA suspended in a non-infectious matrix with carrier DNA, preservatives and stabilizers.