Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ XPert™ Nano Enclosures: 4 ft. Width With Built-in Ionizer
SDP

Catalog No. p-4904424 Shop All Labconco Corporation Products
Change view
Click to view available options
Hertz:
50 Hz
60 Hz
Voltage:
100 V
230 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hertz Voltage
22-167-539 60 Hz 100 V
22-167-541 50 Hz 230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-167-539 Supplier Labconco™ Supplier No. 3887460
Add to Cart
Add to Cart
Request a Quote

Requires

Support base

Keep hazardous powders and particulates contained during procedures

Room air is pulled into the enclosure through the front, flows to the baffle and finally passes through a 99.99% ULPA exhaust filter before returning to the laboratory or cleanroom. Features built-in blower and Guardian™ airflow monitor. With built-in ionizer; separate filtered exhauster is not required.

  • Ergonomic air foil with aerodynamic Clean-Sweep™ airflow openings; side spring pin releases air foil for cleaning
  • Upper dilution air supply and containment sash foil with side-entry air foils
  • Zone-perforated, rear baffle pivots down and removes for cleaning
  • Exhaust ULPA filter, 99.99% efficient on 0.12μm particles; accessible from top for leak scanning of entire surface
  • True bag-in/bag-out filter disposal system (requires certified technician)
  • Built-in blower with dynamically balanced, vibration-isolated motorized impeller and speed control
  • 40% Blower reserve allows speed adjustment (by certified technician) as filter loads
  • Pressure gauge to monitor filter loading
  • Guardian Airflow Monitor continuously monitors airflow; features visual and audible alarms if airflow is outside setpoint range

Construction

  • Choice of interior depth (depending on width selected): 50cm (23.4 in.) or 76cm (30 in.)
  • Type 304 stainless-steel interior sides, baffle, air foil and integral work surface
  • Glacier white and gray, dry powder epoxy-coated aluminum and steel exterior
  • Prewired fluorescent lamp and switches for blower and light
  • 0.6cm (0.25 in.) Thick tempered safety glass front sash
  • 24cm (9.45 in.) Sash opening height on 4 ft. models
  • Ergonomically angled and hinged sash pivots upward, locks in up position; wiping seal contains contaminants
  • Two utility ports for pass-through of electrical cords
Specifications
  • Sash Opening: 9.4 in.
  • At 100fpm Exhaust: 295cfm
  • At 100fpm Noise: 62-66dbA
  • At 80fpm Exhaust: 235cfm
  • At 80fpm Noise: 58-61dbA
  • At 60fpm Exhaust: 175cfm
  • At 60fpm Noise: 49-54dbA

Ideal for nanoparticle manipulation and dry powder chemical handling

Order Info

Optional accessories include canopy connection, remote blowers, ductwork

Compliance

SEFA 1-2002; Modified ASHRAE™ 110-1995; ANSI™ Z9.5-1993; UL 3101-1/61010-1; CAN/CSA C22.2 No. 1010.1; CE Conformity Marking (230V models)

Specifications

Product Type Enclosure
Lighting Type Fluorescent
Material Steel
Certifications/Compliance ANSI Z9.5, CAN/CSA C22.2, CE, ETL, UL 3101-1/61010-1, ASHRAE 110, SEFA 1
Chamber Material Epoxy Coated Aluminum
Color Glacier White, Gray
Depth (English) 29.1 in.
Depth (English) Interior 23.5 in.
Depth (Metric) 74 cm
Depth (Metric) Interior 59.69 cm
Dimensions (L x W x H) 29.1 x 48 x 38.3 in. (73.9 x 122 x 97 cm)
Exhaust ULPA filter
For Use With (Application) Nanoparticle manipulation and dry powder chemical handling
Height (English) 38.3 in.
Height (English) Interior 22.5 in.
Height (English) Sash Opening 9.4 in.
Height (Metric) 97 cm
Height (Metric) Sash Opening 24 cm
Hertz 60 Hz
Length (English) 29.1 in.
Length (Metric) 73.9 cm
Noise Level 58 to 61 dBA
Voltage 100 V
Volume (English) Exhaust 175 to 295 cfm
Width (English) 4 ft.
Width (English) Interior 46.8 in.
Width (Metric) 122 cm
Width (Metric) Interior 118.87 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.