Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ XPert™ Nano Enclosures: 4ft. Width Without Built-in Ionizer
GSA_VASDP

Keep hazardous powders and particulates contained during procedures

$14450.00 - $14680.00

Specifications

Product Type Enclosure
Lighting Type Fluorescent
Material Steel
Chamber Material Epoxy Coated Aluminum
Color Glacier White, Gray
View More Specs

Requires: Support base


Products 2
Catalog Number Mfr. No. Hertz Voltage Price Quantity & Availability  
Catalog Number Mfr. No. Hertz Voltage Price Quantity & Availability  
22-167-538
SDP
View Documents
Labconco™
3887421
50 Hz 230 V
Each for $14,450.00
Only null left
Add to Cart
 
22-167-537
GSA_VASDP
View Documents
Labconco™
3887420
60 Hz 100/115 V
Each for $14,680.00
Only null left
Add to Cart
 
Description

Description

Room air is pulled into the enclosure through the front, flows to the baffle and finally passes through a 99.99% ULPA exhaust filter before returning to the laboratory or cleanroom. Features built-in blower and Guardian™ airflow monitor. With built-in ionizer; separate filtered exhauster is not required.

  • Ergonomic air foil with aerodynamic Clean-Sweep™ airflow openings; side spring pin releases air foil for cleaning
  • Upper dilution air supply and containment sash foil with side-entry air foils
  • Zone-perforated, rear baffle pivots down and removes for cleaning
  • Exhaust ULPA filter, 99.99% efficient on 0.12μm particles; accessible from top for leak scanning of entire surface
  • True bag-in/bag-out filter disposal system (requires certified technician)
  • Built-in blower with dynamically balanced, vibration-isolated motorized impeller and speed control
  • 40% Blower reserve allows speed adjustment (by certified technician) as filter loads
  • Pressure gauge to monitor filter loading
  • Guardian Airflow Monitor continuously monitors airflow; features visual and audible alarms if airflow is outside setpoint range

Construction

  • Choice of interior depth (depending on width selected): 50cm (23.4 in.) or 76cm (30 in.)
  • Type 304 stainless-steel interior sides, baffle, air foil and integral work surface
  • Glacier white and gray, dry powder epoxy-coated aluminum and steel exterior
  • Prewired fluorescent lamp and switches for blower and light
  • 0.6cm (0.25 in.) Thick tempered safety glass front sash
  • 24cm (9.45 in.) Sash opening height on 4 ft. models
  • Ergonomically angled and hinged sash pivots upward, locks in up position; wiping seal contains contaminants
  • Two utility ports for pass-through of electrical cords
Specifications
  • Sash Opening: 9.4 in.
  • At 100fpm Exhaust: 295cfm
  • At 100fpm Noise: 62-66dbA
  • At 80fpm Exhaust: 235cfm
  • At 80fpm Noise: 58-61dbA
  • At 60fpm Exhaust: 175cfm
  • At 60fpm Noise: 49-54dbA

Ideal for nanoparticle manipulation and dry powder chemical handling

Order Info

Optional accessories include canopy connection, remote blowers, ductwork

Compliance

SEFA 1-2002; Modified ASHRAE™ 110-1995; ANSI™ Z9.5-1993; UL 3101-1/61010-1; CAN/CSA C22.2 No. 1010.1; CE Conformity Marking (230V models)

Specifications

Specifications

Enclosure
Steel
Glacier White, Gray
23.5 in.
59.69 cm
ULPA filter
38.3 in.
9.4 in.
24 cm
73.9 cm
175 to 295 cfm
46.8 in.
118.87 cm
Fluorescent
Epoxy Coated Aluminum
29.1 in.
74 cm
29.1 x 48 x 38.3 in. (73.9 x 122 x 97 cm)
Nanoparticle manipulation and dry powder chemical handling
22.5 in.
97 cm
29.1 in.
58 to 61 dBA
48 in.
122 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.