Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ XPert™ Nano Enclosures: 5ft. Width, Extra Deep Without Built-in Ionizer
SDP

Catalog No. p-4904427 Shop All Labconco Corporation Products
Change view
Click to view available options
Frequency:
50 Hz
60 Hz
Voltage:
100/115 V
230 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frequency Voltage
22-167-553 50 Hz 230 V
22-167-552 60 Hz 100/115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-167-553 Supplier Labconco™ Supplier No. 3888521
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Requires

Support base

Keep hazardous powders and particulates contained during procedures

Room air is pulled into the enclosure through the front, flows to the baffle and finally passes through a 99.99% ULPA exhaust filter before returning to the laboratory or cleanroom. Features built-in blower and Guardian™ airflow monitor. With built-in ionizer; separate filtered exhauster is not required.

  • Ergonomic air foil with aerodynamic Clean-Sweep™ airflow openings; side spring pin releases air foil for cleaning
  • Upper dilution air supply and containment sash foil with side-entry air foils
  • Zone-perforated, rear baffle pivots down and removes for cleaning
  • Exhaust ULPA filter, 99.99% efficient on 0.12μm particles; accessible from top for leak scanning of entire surface
  • True bag-in/bag-out filter disposal system (requires certified technician)
  • Built-in blower with dynamically balanced, vibration-isolated motorized impeller and speed control
  • 40% Blower reserve allows speed adjustment (by certified technician) as filter loads
  • Pressure gauge to monitor filter loading
  • Guardian Airflow Monitor continuously monitors airflow; features visual and audible alarms if airflow is outside setpoint range

Construction

  • Choice of interior depth (depending on width selected): 50cm (23.4 in.) or 76cm (30 in.)
  • Type 304 stainless-steel interior sides, baffle, air foil and integral work surface
  • Glacier white and gray, dry powder epoxy-coated aluminum and steel exterior
  • Prewired fluorescent lamp and switches for blower and light
  • 0.6cm (0.25 in.) Thick tempered safety glass front sash
  • Sash opening height on 30cm (11.8 in.) height on 5 ft. models
  • Ergonomically angled and hinged sash pivots upward, locks in up position; wiping seal contains contaminants
  • 5 ft. models incorporate an external gas-assist lift
  • Two utility ports for pass-through of electrical cords
Specifications
  • Sash Opening: 11.8 in.
  • At 100fpm Exhaust: 480cfm
  • At 100fpm Noise: 63-67dbA
  • At 80fpm Exhaust: 385cfm
  • At 80fpm Noise: 58-63dbA
  • At 60fpm Exhaust: 290cfm
  • At 60fpm Noise: 52-56dbA

Ideal for nanoparticle manipulation and dry powder chemical handling

Order Info

Optional accessories include canopy connection, remote blowers, ductwork

Compliance

SEFA 1-2002; Modified ASHRAE™ 110-1995; ANSI™ Z9.5-1993; UL 3101-1/61010-1; CAN/CSA C22.2 No. 1010.1; CE Conformity Marking (230V models)

Specifications

Product Type Enclosure
Lighting Type Fluorescent
Material Steel
Certifications/Compliance CE
Chamber Material Epoxy Coated Aluminum
Color Glacier White, Gray
Depth (English) 29.1 in.
Depth (English) Interior 23.5 in.
Depth (Metric) 74 cm
Depth (Metric) Interior 59.69 cm
Dimensions (L x W x H) 35.6 x 60 x 47.5 in. (90.4 x 152 x 120.65 cm)
Exhaust ULPA filter
For Use With (Application) Nanoparticle manipulation and dry powder chemical handling
Height (English) 47.5 in.
Height (English) Interior 31.8 in.
Height (English) Sash Opening 11.8 in.
Height (Metric) 120.65 cm
Height (Metric) Sash Opening 30 cm
Length (English) 35.6 in.
Length (Metric) 90.4 cm
Noise Level 58 to 63 dBA
Voltage 230 V
Volume (Metric) Exhaust 165.18 to 273.72 L/sec.
Width (English) 60 in.
Width (English) Interior 70.8 in.
Width (Metric) 152.4 cm
Width (Metric) Interior 179.83 cm
Frequency 50 Hz
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.