Learn More
Labconco™ XPert™ Nano Enclosures: 5ft. Width With Built-in Ionizer

Keep hazardous powders and particulates contained during procedures
$25900.00 - $26100.00
Specifications
| Product Type | Enclosure |
|---|---|
| Lighting Type | Fluorescent |
| Material | Steel |
| Chamber Material | Epoxy Coated Aluminum |
| Color | Glacier White, Gray |
Requires: Support base
| Catalog Number | Mfr. No. | Hertz | Voltage | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Hertz | Voltage | Price | Quantity & Availability | ||||
22-167-544
![]() |
Labconco™
3887560 |
60 Hz | 100/115 V |
Each for $25,900.00
|
|
||||
22-167-546
![]() |
Labconco™
3887562 |
50 Hz | 230 V |
Each for $26,100.00
|
|
||||
Description
Room air is pulled into the enclosure through the front, flows to the baffle and finally passes through a 99.99% ULPA exhaust filter before returning to the laboratory or cleanroom. Features built-in blower and Guardian™ airflow monitor. With built-in ionizer; separate filtered exhauster is not required.
- Ergonomic air foil with aerodynamic Clean-Sweep™ airflow openings; side spring pin releases air foil for cleaning
- Upper dilution air supply and containment sash foil with side-entry air foils
- Zone-perforated, rear baffle pivots down and removes for cleaning
- Exhaust ULPA filter, 99.99% efficient on 0.12μm particles; accessible from top for leak scanning of entire surface
- True bag-in/bag-out filter disposal system (requires certified technician)
- Built-in blower with dynamically balanced, vibration-isolated motorized impeller and speed control
- 40% Blower reserve allows speed adjustment (by certified technician) as filter loads
- Pressure gauge to monitor filter loading
- Guardian Airflow Monitor continuously monitors airflow; features visual and audible alarms if airflow is outside setpoint range
Construction
- Choice of interior depth (depending on width selected): 50cm (23.4 in.) or 76cm (30 in.)
- Type 304 stainless-steel interior sides, baffle, air foil and integral work surface
- Glacier white and gray, dry powder epoxy-coated aluminum and steel exterior
- Prewired fluorescent lamp and switches for blower and light
- 0.6cm (0.25 in.) Thick tempered safety glass front sash
- Sash opening height on 30cm (11.8 in.) height on 5 ft. models
- Ergonomically angled and hinged sash pivots upward, locks in up position; wiping seal contains contaminants
- 5 ft. models incorporate an external gas-assist lift
- Two utility ports for pass-through of electrical cords
- Sash Opening: 11.8 in.
- At 100fpm Exhaust: 480cfm
- At 100fpm Noise: 63-67dbA
- At 80fpm Exhaust: 385cfm
- At 80fpm Noise: 58-63dbA
- At 60fpm Exhaust: 290cfm
- At 60fpm Noise: 52-56dbA
Ideal for nanoparticle manipulation and dry powder chemical handling
Order Info
Optional accessories include canopy connection, remote blowers, ductwork
Compliance
SEFA 1-2002; Modified ASHRAE™ 110-1995; ANSI™ Z9.5-1993; UL 3101-1/61010-1; CAN/CSA C22.2 No. 1010.1; CE Conformity Marking (230V models)
Specifications
| Enclosure | |
| Steel | |
| Glacier White, Gray | |
| 23.3 in. | |
| 59.18 cm | |
| ULPA filter | |
| 47.5 in. | |
| 11.8 in. | |
| 30 cm | |
| 73.9 cm | |
| 290 to 480 cfm | |
| 60 in. | |
| 149.35 cm |
| Fluorescent | |
| Epoxy Coated Aluminum | |
| 29.1 in. | |
| 74 cm | |
| 29.1 x 60 x 47.5 in. (73.9 x 152 x 120.65 cm) | |
| Nanoparticle manipulation and dry powder chemical handling | |
| 31.8 in. | |
| 120.65 cm | |
| 29.1 in. | |
| 58 to 63 dBA | |
| 136.86 to 226.53 L/sec. | |
| 58.8 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.