Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Liquid Handling Pack 3 - (Xplorer™ plus pack, Repeater™ E3, Pipette Manager, and More) PROMO

Catalog No. 13684256PM Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
50 to 1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-684-256PM 50 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-684-256PM Supplier Eppendorf™ Supplier No. 2231001062

missing translation for 'validMainframeMsg'

Save up to 40% on qualifying Eppendorf™ pipettes and liquid handling product packages.

Promo: Save up to 40% on qualifying Eppendorf™ pipettes and liquid handling products:

NOTE: Discounts taken off list price. Prices shown reflect promotional prices.

Liquid Handling Package 3 Includes:

  • Eppendorf™ Xplorer™ Plus electronicsingle-channel pipettes (0.5 - 10 μL, 5 - 100 μL, 50 - 1,000 μL)
  • Repeater E3 electronic dispenser (Cat. No. 13-683-550)
  • Easypet™ 3 Pipette Controller
  • Pipette Manager (1 module) (Cat. No. 02-911-585)
  • Charging Carousel 2

Xplorer™ plus single-channel pipettes Features

Intuitive operating system and design for simplicity, precision and reproducibility.

  • Clearly arranged color display; no sub-menus; help function
  • Innovative rocker: When the tip is ejected, the piston automatically returns to zero position
  • Additional applications include aspirating, diluting, sequential dispensing and reverse pipetting
  • Programming: One-time programming of sequences for easier handling of complex processes
  • Individual fixed volume setting: Desired volume can be saved
  • Individual speed setting: Sets parameters to your individual working speed

Repeater™ E3 electronic dispenser (Cat. No. 13-683-550) Features

  • Automatic Combitip recognition: Eliminates time consuming volume calculations, avoiding incorrect dispensing volumes
  • Speed Adjustment: Program aspiration and dispensing speed for the utmost in precision and accuracy while also preventing splashing
  • Repetitive dispensing modes: Decrease the number of movements for the user by allowing them to fill the tip once and dispense up to 100 times without a refill
  • Wide dispensing range: 1 μL to 50 mL provides up to 5,000 dispensing volumes with increments as low as 100 nanoliters
  • Motor driven function reduces the human error to a minimum and reduces risk of repetitive strain injuries (RSI)
  • Large illuminated color display reduces eye strain when reading

Easypet™ 3 pipette controller (Cat. No. 12-654-105) Features

  • 0.1 to 100 mL
  • Vibrant backlit LEDs provide optical feedback of the remaining battery life
  • Lithium polymer rechargeable battery offers long cordless runtime
  • Operation while recharging is possible
  • Autoclavable pipette adapter for sterile applications
  • Quick release of aspirating cone for easy exchange of membrane filters

Pipette Manager (Cat. No. 02-911-585) Features

  • Connected electronic pipette system to remotely set up electronic pipettes, ease handling of challenging liquids and digitally document pipetting steps
  • Easily enter pipetting volumes and other parameters using the touch screen. Settings are immediately transferred to connected electronic pipettes, allowing you to start working right away
  • The software-embedded Liquid Manager sets optimal pipetting settings (e.g., pre-wetting steps, speed) when working with viscous, volatile and other challenging liquid types
  • Choose from predefined Eppendorf liquid types (e.g., ethanol or enzymes/antibodies) or create your own
  • Automatically document pipetting activities via Pipetting Records and easily export your recordings as comprehensive PDF document via USB
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±3% at ±3 μL/100 μL, ±1% at ±5 μL/500 μL, ±0.6% at ±6μL
Battery Type Lithium-ion
Compatible Tips 1000 μL epT.I.P.S™ pipette tip
Color Blue
For Use With epT.I.P.S. Pipette Tip (1000 μL)
Number of Channels 1
Product Line Xplorer
Type Electronic Pipette
Volume (Metric) 50 to 1000 μL
Finger Style Ergonomic
For Use With (Equipment) 1000 μL epT.I.P.S™ pipette tip
Includes 3x Eppendorf Xplorer™ single-channel pipettes (0.5 to 10 μL, 5 to 100 μL, 50 to 1000 μL), charging carousel, Repeater™ E3 electronic dispenser, Easypet™ 3 pipette controller, Pipette Manager
Increments 1 μL
Multilingual User Interface Yes
Number of Speeds 8
Temperature (English) Operating 41°F to 104°F
Temperature (Metric) Operating 5°C to 40°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.