Learn More
Invitrogen™ XPR1 Polyclonal Antibody

Description
PA1-23547 detects Xenotropic & Polytropic Retrovirus Rec. in human samples.
Specifications
Specifications
| Antigen | XPR1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | Xpr1 |
| Gene Accession No. | Q9QZ70, Q9UBH6, Q9Z0U0 |
| Gene Alias | IBGC6; Protein SYG1 homolog; receptor for MCF virus 1; Rmc1; Rmc-1; RP11-46A10.1; suppressor of yeast G>a> deletion; susceptibility to xenotropic virus; Sxv; Syg1; X3; xenotropic and polytropic murine leukemia virus receptor; xenotropic and polytropic murine leukemia virus receptor X3; Xenotropic and polytropic retrovirus receptor 1; xenotropic and polytropic retrovirus receptor 1 homolog; XPR1; XR; X-receptor |
| Gene Symbols | Xpr1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.