Learn More
Diagnostic Biosystems XRCC1 XRCC1
Supplier:  Diagnostic Biosystems MOB32505
Encompass_Preferred
This antibody is specific to an 85 kDa protein known as XRCC1. X-ray repair cross complementing 1 (XRCC1) protein plays a role in excision repair of DNA after ionizing irradiation. XRCC1 binds to DNA ligase III through the Cterminal 96 amino acids and to DNA polymerase β through the N-terminal half. In testis XRCC1 is expressed at high levels.
Catalog No. 50-238-5485
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.