Learn More
Waters Corp XSelect CSH C18 Prep Guard Cartridge, 130Å, 5 µm, 30 mm X 10 mm, 1/pk
Supplier:  Waters Corp 186006899
Encompass_Preferred
30x10mm Prep Guard Cartridge together with the 30x10mm Prep Guard Holder (part number 186006912) is designed to protect both HPLC and SFC 30 mm ID Prep OBD columns. Placing this guard assembly ahead of the prep column will provide longer column lifetimes and reduce purification costs.
Catalog No. 50-596-874
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.